Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
4 ea | Warenkorb auf Verfügbarkeit prüfen | € 177,00 |
Über diesen Artikel
€ 177,00
material
flat bottom bottle, polypropylene copolymer (with sealing closure), round bottle, translucent bottle
joint
Joints threaded neck (63)
feature
RCF 13,700 × g, with cap
packaging
pack of 4 ea
manufacturer/tradename
Nalgene® 3141-0500
parameter
13700 × g max. RCF
bottle diam. × L
69.5 mm × 159.3 mm
nominal capacity
500 mL
General description
Features and Benefits
- Sturdy polypropylene copolymer (PPCO).
- Designed for low- to moderate-centrifugation speed, large-volume, biological and chemical samples.
- Ideal for use in cell culture harvesting and protein biochemistry applications, including ammonium sulfate precipitations.
- Features a leakproof sealing closure for sample protection.
- Autoclave safe.
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description nominal capacity 500 mL | description nominal capacity 250 mL | description nominal capacity 500 mL | description nominal capacity 250 mL |
| feature RCF 13,700 × g, with cap | feature with cap, RCF 27,500 × g | feature RCF 13,700 × g, with cap | feature RCF 27,500 × g, with cap |
| manufacturer/tradename Nalgene® 3141-0500 | manufacturer/tradename Nalgene® 3141-0250 | manufacturer/tradename Nalgene® 3140-0500 | manufacturer/tradename Nalgene® 3140-0250 |
| joint Joints threaded neck (63) | joint Joints threaded neck (58) | joint Joints threaded neck (52.6) | joint Joints threaded neck (46.5) |
| material translucent bottle, polypropylene copolymer (with sealing closure), flat bottom bottle, round bottle | material polypropylene copolymer (with sealing closure), translucent bottle, flat bottom bottle | material round bottle, transparent polycarbonate (with sealing closure), screw top cap, transparent bottle, polypropylene bottle | material screw top cap, polypropylene bottle, transparent bottle, transparent polycarbonate (with sealing closure), round bottle |
| packaging pack of 4 ea | packaging pack of 4 ea | packaging case of 24, pack of 4 | packaging pack of 4 |
| parameter 13700 × g max. RCF | parameter 27500 × g max. RCF | parameter 13700 × g max. RCF | parameter 27500 × g max. RCF |
Still not finding the right product?
Explore all of our products under Nalgene® centrifuge bottles, wide-mouth with sealing caps Style 3141
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?