Fortfahren mit
CLS3788
Corning® 96-Well transparente Mikrotiterplatte aus Polystyrol
round bottom clear, polystyrene, bag of 20 ×, sterile, lid
Synonym(e):
96 multiwell plates, 96 well microplates, 96 well plates
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
100 ea
Warenkorb auf Verfügbarkeit prüfen
€ 377,00
Über diesen Artikel
NACRES:
NB.13
UNSPSC Code:
41121800
Material:
clear bottom, polystyrene , round bottom clear
Size:
96 wells
€ 377,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
clear bottom, polystyrene , round bottom clear
sterility
sterile
feature
lid, skirt
manufacturer/tradename
Corning 3788
packaging
case of 100, bag of 20 ×
size
96 wells
well volume
330 μL
working volume
75-200 μL
suitability
suitable for (cell cuture)
binding type
non-treated surface
General description
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
  • Untreated microplates have a binding capacity of approximately 100 to 200 ng IgG/cm2
  • Total volume is 330 μL; recommended working volume of up to 75 to 200 μL
  • Conforms to standard microplate footprint and dimensions
Note: Total well volume is 330 μL
Legal Information
Corning is a registered trademark of Corning, Inc.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
CLS3370CLS3795CLS3898
description
round bottom clear, polystyrene, bag of 20 ×, sterile, lid
description
flat bottom clear, polystyrene, sterile, lid, case of 100, pack of 20
description
round bottom clear, polystyrene, bag of 25, sterile, lid: no
description
V-bottom clear, polystyrene, bag of 25, non-sterile, lid: no
suitability
suitable for (cell cuture)
suitability
suitable for (EIA/RIA; immunoassays)
suitability
suitable for (cell cuture)
suitability
suitable for (cell cuture)
size
96 wells
size
96 wells
size
96 wells
size
96 wells
material
round bottom clear , clear bottom, polystyrene
material
flat bottom clear , polystyrene , round wells
material
round bottom clear , round wells, polystyrene , clear wells
material
V-bottom clear , clear bottom, polystyrene
packaging
bag of 20 × , case of 100
packaging
case of 100, pack of 20
packaging
bag of 25
packaging
bag of 25
feature
lid, skirt
feature
lid, skirt (skirt)
feature
lid: no, skirt
feature
skirt, lid: no
well volume
330 μL
well volume
360 μL
well volume
330 μL
well volume
-
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)