Merck
BAF134880050
Bel-Art® Write-on Label Tape
roll size 40 yard, orange, width 1/2 in., pk of 6
Synonym(e):
labelling tape, moisture resistant label tape, solvent resistant label tape
Größe auswählen
6 EA
€ 87,00
€ 87,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
6 EA
€ 87,00
About This Item
UNSPSC-Code:
55121611
NACRES:
NB.35
€ 87,00
Check Cart for Availability
Materialien
orange
Verpackung
pk of 6
Hersteller/Markenname
Bel-Art F13488-0050
Walzengröße
40 yard
Breite
1/2 in.
12.7 mm
Allgemeine Beschreibung
Select from six tape colors that correspond with familiar hazard color codes to highlight safety information written on the labels. Use yellow for chemical reactivity, green for safety labels, blue indicates health hazards, red alerts to flammability; orange indicates a biohazard and white can describe a specific hazard. The tape is custom manufactured from proprietary materials that resist moisture and most solvents. Will adhere to PTFE fluoropolymer resin covered materials and any clean surface. A maximum temperature of 247 degrees C for 30 minutes can be endured.
Leistungsmerkmale und Vorteile
  • No sticky residue lingers when tape is stripped off
  • Marks made with pencil, ball-point pen or solvent ink marker survive most laboratory conditions
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Write-on is a trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)