Merck
AXYPCRTS
CyclerSeal sealing film
size 60 μm, polypropylene, pkg of 5x100sheets/cs
Synonym(e):
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate
About This Item
UNSPSC-Code:
41122410
NACRES:
NB.22
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
polypropylene
Sterilität
non-sterile
Hersteller/Markenname
Axygen Inc PCR-TS
Corning
Verpackung
pkg of 5x100sheets/cs
Größe
60 μm
Allgemeine Beschreibung
  • Axygen® CyclerSeal sealing film is made of high-quality, temperature-resistant polypropylene material.
  • Reduces well-to-well contamination and cross-over in PCR application when used in conjunction with Axygen′s compression mat.
  • Ideal for PCR, Real-time PCR, storage, ELISA and cell culture applications.
  • The sealing film offers a tight seal across the entire plate and ensures optimal sample protection during transport or storage.
Rechtliche Hinweise
Axygen is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)