Merck
AXYP96450VC
V-bottom assay plate
size 96 wells, maximum volume 500 μL, V-bottom clear polypropylene wells, pkg of 10x5plates/cs
Synonym(e):
V-bottom well plate
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
Materialien
V-bottom clear polypropylene wells
clear plate
Sterilität
non-sterile
Leistungsmerkmale
lid: no
skirt
Hersteller/Markenname
Axygen Inc P-96-450V-C
Corning
Verpackung
pkg of 10x5plates/cs
Maximales Volumen
500 μL
Größe
96 wells
Arbeitsvolumen
450 μL
Bindungsart
non-treated surface