Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 184,00 |
Über diesen Artikel
€ 184,00
material
borosilicate glass , round bottom flask, three-neck (Vertical), three-neck
joint
joint: ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
500 mL
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description vertical, capacity 500 mL, joint: ST/NS 24/40 | description vertical, capacity 250 mL, joint: ST/NS 24/40 | description angled, capacity 500 mL, joint: ST/NS 24/40 | description vertical, capacity 1,000 mL, joint: ST/NS 24/40 |
| manufacturer/tradename Sigma-Aldrich | manufacturer/tradename Sigma-Aldrich | manufacturer/tradename Sigma-Aldrich | manufacturer/tradename Sigma-Aldrich |
| capacity 500 mL | capacity 250 mL | capacity 500 mL | capacity 1,000 mL |
| joint joint: ST/NS24/40 | joint joint: ST/NS24/40 | joint joint: ST/NS24/40 | joint joint: ST/NS24/40 |
| material three-neck , three-neck (Vertical), borosilicate glass , round bottom flask | material round bottom flask, borosilicate glass , three-neck , three-neck (Vertical) | material three angled neck , round bottom flask, glass (Clear) | material three-neck (Vertical), round bottom flask, borosilicate glass |
| packaging pack of 1 | packaging pack of 1 | packaging case of 1 | packaging pack of 1 |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
Still not finding the right product?
Explore all of our products under Aldrich® Dreihals-Rundkolben
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?