Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 195,00 |
Über diesen Artikel
€ 195,00
material
borosilicate glass , round bottom flask, three-neck (Vertical), three-neck
joint
joint: ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
1,000 mL
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description vertical, capacity 1,000 mL, joint: ST/NS 24/40 | description vertical, capacity 1,000 mL, joint: ST/NS 29/32 | description angled, capacity 1,000 mL, joint: ST/NS 29/32 | description vertical, capacity 100 mL, joint: ST/NS 14/20 |
| capacity 1,000 mL | capacity 1,000 mL | capacity 1,000 mL | capacity 100 mL |
| manufacturer/tradename Sigma-Aldrich | manufacturer/tradename Sigma-Aldrich | manufacturer/tradename Sigma-Aldrich | manufacturer/tradename Sigma-Aldrich |
| joint joint: ST/NS24/40 | joint joint: ST/NS29/32 | joint joint: ST/NS29/32 | joint joint: ST/NS14/20 |
| material borosilicate glass , three-neck (Vertical), round bottom flask | material borosilicate glass , round bottom flask, three-neck (Vertical) | material borosilicate glass , round bottom flask, three angled neck , three-neck | material borosilicate glass , round bottom flask, three-neck (Vertical) |
| packaging pack of 1 | packaging pack of 1 | packaging pack of 1 | packaging pack of 1 |
| feature closure type Standard Taper Joint | feature closure type Standard Taper Joint | feature closure type Standard Taper Joint | feature closure type Standard Taper Joint |
Still not finding the right product?
Explore all of our products under Aldrich® Dreihals-Rundkolben
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?