Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
glass body
needle size
(not included)
feature
without needle Luer tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81501
needle L
, n/a
volume
5 mL
application(s)
general analytical
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description 1005LT, volume 5 mL, needle size (not included) | description 1001LT, volume 1 mL, needle size (not included) | description 1002LT, volume 2.5 mL, needle size (not included) | description 1010LT, volume 10 mL, needle size (not included) |
| material glass body | material glass body | material glass body | material glass body |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| feature without needle Luer tip | feature without needle Luer tip | feature without needle Luer tip | feature without needle Luer tip |
| volume 5 mL | volume 1 mL | volume 2.5 mL | volume 10 mL |
| manufacturer/tradename Hamilton 81501 | manufacturer/tradename Hamilton 81301 | manufacturer/tradename Hamilton 81401 | manufacturer/tradename Hamilton 81601 |
| needle L , n/a | needle L , n/a | needle L , n/a | needle L , n/a |
Still not finding the right product?
Explore all of our products under Hamilton® Spritze, Serie 1000 GASTIGHT®, Luer-Spitze
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?