Direkt zum Inhalt
Merck
Alle Fotos(4)

Wichtige Dokumente

MABN2637

Sigma-Aldrich

Anti-AIDA1 Antibody, clone 2B22

Anmeldenzur Ansicht organisationsspezifischer und vertraglich vereinbarter Preise


About This Item

UNSPSC-Code:
12352203
NACRES:
NA.41

Biologische Quelle

mouse

Qualitätsniveau

Konjugat

unconjugated

Antikörperform

purified antibody

Antikörper-Produkttyp

primary antibodies

Klon

2B22, monoclonal

Mol-Gew.

calculated mol wt 139.21, 48.37, and 57.81 kDa
observed mol wt ~50 and 55 kDa

Aufgereinigt durch

using protein G

Speziesreaktivität

rat

Speziesreaktivität (Voraussage durch Homologie)

canine, feline

Verpackung

antibody small pack of 100 μg

Isotyp

IgG2κ

Epitopsequenz

C-terminal

UniProt-Hinterlegungsnummer

Versandbedingung

2-8°C

Posttranslationale Modifikation Target

unmodified

Allgemeine Beschreibung

Ankyrin repeat and sterile alpha motif domain-containing protein 1B (UniProt: P0C6S7; also known as Amyloid-beta protein intracellular domain-associated protein 1, AIDA-1, E2A-PBX1-associated protein, EB-1) is encoded by the Anks1b gene (Gene ID: 314721) in rat. AIDA-1 is an adaptor protein that has been implicated in Cajal body and nucleolar function. It is expressed throughout the brain and is highly expressed in the hippocampus. It is one of the most abundant proteins at postsynaptic densities where it is reported to bind to PSD95 and associates with NMDA receptors (NMDARs). In the brain it interacts with amyloid b precursor protein and plays a role in brain development. AIDA-1 preferentially associates with GluN2B-containing NMDARs and plays a role in their transport from the endoplasmic reticulum (ER). Loss of AIDA-1 results in the accumulation of GluN2B in the ER and decreases GluN2B and increases GluN2A distribution at the synapse. However, loss of AIDA-1 does not affect AMPAR-mediated synaptic transmission or the probability of glutamate release. Mutations in Anks1b gene are known to affect GluN2B function and are linked to the pathogenesis of neuropsychiatric disorders. (Ref.: Carbonell, AU., et al., (2019). Nat. Commun. 10(1); 3529; Tindi, JO., et al. (2015). J. Neurosci. 35(24); 8986- 8996).

Spezifität

Clone 2B22 is a is a mouse monoclonal antibody that detects AIDA-1. It targets an epitope within 10 amino acids from the C-terminal region.

Immunogen

A linear peptide corresponding to 10 amino acids from the C-terminal region of rat Ankyrin repeat and sterile alpha motif domain-containing protein 1B (AIDA-1).

Anwendung

Quality Control Testing

Evaluated by Western Blotting in Rat brain tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected AIDA1 in Rat brain tissue lysate.

Tested Applications


Western Blotting Analysis: A representative lot detected AIDA1 in Western Blotting applications (Tindi, J.O., et al. (2015). J Neurosci. 35(24): 8986-96).

Immunofluorescence Analysis: A representative lot detected AIDA1 in Immunofluorescence applications (Tindi, J.O., et al. (2015). J Neurosci. 35(24): 8986-96).

Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected AIDA1 in rat cerebral cortex, hippocampus, and cerebellum tissue sections.

Immunoprecipitation Analysis: A representative lot immunoprecipitated AIDA1 in Immunoprecipitation applications (Carbonell, A.U., et al. (2019). Nat Commun.10(1): 3529).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-AIDA1, clone 2B22, Cat. No. MABN2637, is a mouse monoclonal antibody that detects AIDA1/ANKS1B and is tested for use in Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.

Physikalische Form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Lagerung und Haltbarkeit

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Sonstige Hinweise

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Haftungsausschluss

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Sie haben nicht das passende Produkt gefunden?  

Probieren Sie unser Produkt-Auswahlhilfe. aus.

Lagerklassenschlüssel

12 - Non Combustible Liquids

WGK

WGK 1

Flammpunkt (°F)

Not applicable

Flammpunkt (°C)

Not applicable


Analysenzertifikate (COA)

Suchen Sie nach Analysenzertifikate (COA), indem Sie die Lot-/Chargennummer des Produkts eingeben. Lot- und Chargennummern sind auf dem Produktetikett hinter den Wörtern ‘Lot’ oder ‘Batch’ (Lot oder Charge) zu finden.

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen

Unser Team von Wissenschaftlern verfügt über Erfahrung in allen Forschungsbereichen einschließlich Life Science, Materialwissenschaften, chemischer Synthese, Chromatographie, Analytik und vielen mehr..

Setzen Sie sich mit dem technischen Dienst in Verbindung.