96320250
Vantage™ L Laboratory Columns
Vantage™ L Laboratory Column VL 32 x 250
Synonym(e):
Vantage L Laboratory Column VL 32 x 250
About This Item
Empfohlene Produkte
Qualitätsniveau
Produktlinie
Vantage
Parameter
operating temperature at 2-40 °C
6.9 bar max. inlet pressure (100 psi)
7 bar max. pressure (100 psid)
Methode(n)
protein purification: suitable
Länge
250 mm
Bereich
8 cm2 , cross-sectional
Bett H
0-250 mm
Bettvolumen
0-200 mL
Durchmesser
32-32.1 mm