Merck
Z766852
Microflex NeoPro® EC powder-free chloroprene gloves
size M
Synonym(e):
disposable glove
About This Item
UNSPSC-Code:
46181504
NACRES:
NB.35
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
(chloroprene gloves)
green
Größe
M
Beschreibung
ambidexterous
textured fingers
Leistungsmerkmale
CE compliant
disposable
latex free
powder-free
Verpackung
pack of 50 ea (boxes of 50)
Hersteller/Markenname
Microflex NEC-288-M
Länge
11.8 in. , cuff
Dicke
6.3 mil , palm
8.3 mil , fingers
Allgemeine Beschreibung
Featureing an extended cuff, NeoPro® EC is the longest and thickest non-latex glove produced by Microflex. It uses a polychloroprene formulation to provide a secure grip along with exceptional protection from biological and chemical hazards inwet or dry conditions.
Rechtliche Hinweise
NeoPro is a registered trademark of CIT Healthcare LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)