Merck
Z748293
Sigma-Aldrich
Aldrich® Kugelrohr glassware set
25 mL and 100 mL load and ball-tube flasks with straight tube, joint: ST/NS 14/20
Synonym(e):
distillation apparatus, distilling apparatus, fractional distillation apparatus, glass distillation apparatus, kugelrohr apparatus, kugelrohr distillation, short path distillation apparatus, vacuum distillation apparatus
Größe auswählen
1 SET
€ 335,00
€ 335,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 SET
€ 335,00
About This Item
UNSPSC-Code:
41104812
NACRES:
NB.43
€ 335,00
Check Cart for Availability
Verbindung
joint: ST/NS14/20
Verpackung
set of 1 pkg
Verwandte Kategorien
Allgemeine Beschreibung
The set contains: two load flasks (25 and 100 mL), two ball tube (receiving) flasks (25 and 100 mL) and a straight tube.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Questions
  1. What are the dimensions of Z748293, Aldrich® Kugelrohr glassware set?
    1 answer
    1. The Aldrich Kugelrohr glassware set, product Z748293, is a 25 mL set-up consisting of a load flask, receiver flask and straight tube.  It has an assembled length of 490 mm, and approximately 195 mm extends from the side of the oven.The 100 mL set-up has an assembled length of 515 mm approximately, and 205 mm extends from the side of the oven.