Merck
Z743026
Julabo® CORIO CD heating circulator with open bath
CD-B39, capacity 35 L (41 L), temperature 20-150 °C, stainless steel bath, AC/DC input 230 V AC, Schuko plug
Größe auswählen
1 EA
€ 3.300,00
€ 3.300,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 3.300,00
About This Item
UNSPSC-Code:
41103706
NACRES:
NB.77
€ 3.300,00
Check Cart for Availability
Materialien
stainless steel bath
Verpackung
pack of 1 ea
Hersteller/Markenname
Julabo 9012439
Temperaturmessbereich
20-150 °C
Kapazität
35 L (41 L)
Außenmaß B × L × H
54 cm × 34 cm × 57 cm
Öffnung B × L × H
33 cm × 30 cm × 30 cm
Allgemeine Beschreibung
The Open Heating Bath Circulators of the new CORIO series feature high-quality bath tanks made of stainless steel and pump connections.
Your advantages
  • Models for internal and external applications
  • Bright, white, easy to read display
  • Very quiet
  • Easy pump change-over between internal and external circulation
  • USB connection
  • High-quality bath tanks made of stainless steel
  • Integrated drain screw (except B5)
Cooling coil and bath cover available as optional accessories
Rechtliche Hinweise
CORIO is a trademark of Julabo GmbH
Julabo is a registered trademark of Julabo USA, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)