Merck
Z742448
MAGic Clamp magnetic clamp
holds 3000 mL Erlenmeyer flask
Synonym(e):
flask clamps, magnetic clamps
Größe auswählen
1 EA
€ 121,00
€ 121,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 121,00
About This Item
UNSPSC-Code:
41122413
NACRES:
NB.73
€ 121,00
Check Cart for Availability
Leistungsmerkmale
holds 3000 mL Erlenmeyer flask
max 4x3000ml flask
Verpackung
pkg of 1
Hersteller/Markenname
Benchmark Scientific H1000-MR-3000
Allgemeine Beschreibung
The convenient MAGic Clam platform for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes.
designed to be used with Benchmark Scientific H1010 shaking incubators and OrbiShaker XL orbital shakers
Rechtliche Hinweise
MAGic Clamp is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)