Merck
Z742306
Horizontal rotisserie for RotoBot Rotating Mixers
holds 6 x 15 mL tubes, pkg of 1 ea
Größe auswählen
1 EA
€ 78,60
€ 78,60
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 78,60
About This Item
UNSPSC-Code:
41103801
NACRES:
NB.73
€ 78,60
Check Cart for Availability
Leistungsmerkmale
holds 6 x 15 mL tubes
Verpackung
pkg of 1 ea
Hersteller/Markenname
Benchmark Scientific R4040-HZ15
Röhrchendurchm.
16-17 mm
Allgemeine Beschreibung
For higher capacity applications, these holders can be purchased as accessories for the Rotobot and Rotobot Mini rotating mixers. In addition, horizontal holders are available for applications such as Western blotting and hybridization. The unique, sliding attachment allows for quick and easy changing of tube holders.
Rechtliche Hinweise
Rotobot is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)