Merck
Z674567
GyroMini nutating mixer
AC/DC input 230 V AC, Schuko plug
Synonym(e):
Nutating mixer
Größe auswählen
1 EA
€ 385,00
€ 385,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 385,00
About This Item
UNSPSC-Code:
41103801
NACRES:
NB.73
€ 385,00
Check Cart for Availability
Hersteller/Markenname
Labnet S0500-230V-EU
Allgemeine Beschreibung
  • GyroMini nutating mixer is designed for maintaining the uniform suspension of blood components in blood collection tubes without foaming.
  • Ideal for mixing large strands of DNA into solution in micro tubes or mixing blood into a buffer in large centrifuge tubes.
  • Small foot print means less bench space needed
  • Gyratory action provides gentle, thorough mixing
  • Ideal for mixing blood samples in centrifuge tubes
  • Fixed speed and pitch provide optimum agitation
  • Safe for cold room use
The GyroMini is supplied with a dimpled mat for holding tubes firmly in place.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
GyroMini is a trademark of Labnet Intl., Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)