Merck
Z741340
MAGic Clamp universal platform for flasks and tube racks
L × W 16 in. × 16.5 in.
Größe auswählen
1 EA
€ 445,00
€ 445,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 445,00
About This Item
UNSPSC-Code:
41122413
NACRES:
NB.77
€ 445,00
Check Cart for Availability
Hersteller/Markenname
Benchmark Scientific BT4010-MR
L × B
16 in. × 16.5 in.
Allgemeine Beschreibung
The convenient MAGic Clamp platform can be used for both Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes.
Rechtliche Hinweise
MAGic Clamp is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)