Merck
Z740680
Sigma-Aldrich
Aldrich® Essentials filter flask, with side arm
capacity 250 mL
Größe auswählen
12 EA
€ 220,00
€ 220,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
12 EA
€ 220,00
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
€ 220,00
Check Cart for Availability
Materialien
borosilicate glass flask
flat bottom flask
Verbindung
joint stopper6
Verpackung
pkg of 12 ea
Kapazität
250 mL
Allgemeine Beschreibung
Clear borosilicate glass Erlenmeyer style flask with side arm for connecting to vacuum or aspirator line. Used for filtration with Buchner funnels, Gooch crucibles, etc. Beaded rim for rubber gromets or filter adapters or rubber stoppers.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Questions
  1. Which type of flask is suitable for a #6 stopper?
    1 answer
    1. The flask Z740680 is not sized for a #6 stopper, as its opening is too wide, causing the stopper to fall through. However, the item CLS70340250 does fit a #6 stopper.