Merck
Z677345
Dermatril LR nitrile gloves
size XL
Synonym(e):
disposable glove
About This Item
UNSPSC-Code:
46181504
NACRES:
NB.35
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
blue gloves
blue nitrile
Größe
XL
Sterilität
non-sterile
Leistungsmerkmale
CE compliant
latex free
powder-free
Verpackung
pack of 50 ea (boxes of 50)
Hersteller/Markenname
KCL 742 -10
KCL 742
L × Dicke
270 mm × 0.11 mm
Allgemeine Beschreibung
Dermatril LR is an extended cuff, blue nitrile, powder-free glove providing a high level of protection against chemical and bacteriological contamination. The Dermatril LR disposable glove also displays very good physical durability in comparison to conventional disposable gloves.
All Dermatril LR disposable gloves are INDIVIDUALLY marked with the appropriate standards mark applied on the inside using a food-friendly dye.
  • Cleanroom class 100
  • Anti-static
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea
  • Extended cuff
  • Roughened fingertips for better grip
  • Rolled cuff edge for easier donning
  • CE compliant
  • Meet standards: EN 374 (cat. III), EN 388 (cat. II), EN 455, EN 61340-5-1, ASTM 1671:2001, US-FED-ST 209E
Rechtliche Hinweise
Dermatril is a trademark of KCL GmbH
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)