Merck
Z567167
Sigma-Aldrich
VerSA-Flow can/drum adapter
57 mm buttress thread to 2 in. NPT
Größe auswählen
1 EA
€ 29,10
€ 29,10
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 29,10
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.43
€ 29,10
Check Cart for Availability
Allgemeine Beschreibung
Adapts equipment with standard tapered 2 inch NPT thread such as VerSA-Flow dispensers, pumps, faucets and other hardware to plastic or composite drums with 57 mm Buttress threads.
High density polyethylene construction.
Rechtliche Hinweise
VerSA-Flow is a trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)