Merck
Z562084
Platinum crucible lid
fits 25 mL crucible
Synonym(e):
crucibles
Größe auswählen
1 EA
€ 869,00
€ 869,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 869,00
About This Item
UNSPSC-Code:
41122200
NACRES:
NB.43
€ 869,00
Check Cart for Availability
Hersteller/Markenname
Birmingham metals
Gewicht
7.2 g
Allgemeine Beschreibung
Platinum labware has excellent resistance to acids such as acetic, hydrochloric, hydrofluoric, hydroiodic, nitric, selenic, and sulfuric, and to oxidation at high temperatures. Crucible lids of platinum can be cleaned by boiling in a suitable solvent.
For the removal of substances which have alloyed with the surface, scour with alumina powder.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)