Z551872
Aldrich® check-valve bubbler with SafetyBarb® hose connectors
T-inlet tube, side outlet
Synonym(e):
bubblers, scientific bubblers
Empfohlene Produkte
Kapazität
15 mL
Verwandte Kategorien
Allgemeine Beschreibung
Check-valve ball prevents oil from being drawn into the system. Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany