Merck
SLW4510/02
Pyrex® Quickfit(R) Polypropylene screwthread Connector
13 mm Screwthread Inlet and Outlet
Synonym(e):
Pyrex, SciLabware 4510/02
Größe auswählen
10 EA
€ 94,10
€ 94,10
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EA
€ 94,10
About This Item
UNSPSC-Code:
12171500
NACRES:
NB.35
€ 94,10
Check Cart for Availability
Materialien
Viton® O-ring
polypropylene
Hersteller/Markenname
4510/02
SciLabware
Allgemeine Beschreibung
These screwthread hose connectors are only for use with the QUICKFIT® and PYREX® glassware products. They do not fit DURAN® glass threads.
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
PYREX is a registered trademark of Corning, Inc.
QUICKFIT is a registered trademark of DWK Life Sciences
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)