Merck
Z549304
Sigma-Aldrich
Aldrich® anticlimb splash-guard adapter with fritted disc
female joint: ST/NS 29/32, male joint: ST/NS 14/20, capacity 100 mL
Größe auswählen
1 EA
€ 189,00
€ 189,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 189,00
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
€ 189,00
Check Cart for Availability
Verbindung
female joint: ST/NS29/32
male joint: ST/NS14/20
Kapazität
100 mL
Porengröße
70-100 μm porosity
Verwandte Kategorien
Allgemeine Beschreibung
These traps greatly reduce carry-over from foaming or bumping. Because the venting hole is located at the top, the foam can be prevented from climbing higher into the condenser. The fritted disc prevents particulates from entering the condenser.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)