Merck
Z281239
Antistatik-Bürste
ideal for balance and lens cleaning
Synonym(e):
Anti-Static Cleaning Brush
Größe auswählen
1 EA
€ 82,20
€ 82,20
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 82,20
About This Item
UNSPSC-Code:
47131605
NACRES:
NB.35
€ 82,20
Check Cart for Availability
B × L
66 mm × 63 mm
Allgemeine Beschreibung
Ziegenhaar-Bürste. Ideal zur Reinigung von Waagen und Linsen.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Questions
  1. Hi, Could you advise what is the brush wire made of? * Z281239 Anti-static brush I need a Conductive RUBBER Brush. Thank you
    1 answer
    1. The unique blend of soft natural hairs and conductive synthetic fibers plus the conductive nature of the construction result in effective static dissipation that removes static as it cleans.