Merck
Z248096
Sigma-Aldrich
Handbetriebene Nalgene® Vakuumpumpe
PVC body
Synonym(e):
hand vacuum pump, liquid dispensing device
Größe auswählen
1 EA
€ 322,00
€ 322,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 322,00
About This Item
UNSPSC-Code:
41105100
NACRES:
NB.79
€ 322,00
Check Cart for Availability
Materialien
PVC body
Verpackung
pack of 1 ea
Hersteller/Markenname
Nalgene 6132-0010
Verwandte Kategorien
Allgemeine Beschreibung
Pump with vacuum gauge attains and holds vacuum of 125 mm Hg (absolute). Vacuum trigger release requires only one hand to operate. Adjustable vacuum release rate controlled by lever on pump. Full blank port pressure obtained in two strokes.
Removable cover on exhaust port and trigger releases vacuum without disconnecting pump from line. Gauge monitors vacuum in cm and in. Hg.
Includes: 2 ft of 1/4 in. I.D. Nalgene 180 tubing.
Use for siphoning, filtration, liquid transfer, bleeding fluid lines, checking for equipment leaks, and adjusting vacuum-operated devices. When transferring liquids, use air trap to prevent liquid from entering pump.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)