Merck
Z203610
Sigma-Aldrich
Schwarze Kunststoffkappen
Solid-top, PTFE-faced rubber liner, thread for 15-415
Synonym(e):
240463
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.11
Verpackung
pkg of 200 ea
Hersteller/Markenname
WHEATON®
Fitting
thread for 15-415
Allgemeine Beschreibung
These autoclavable caps are designed to ensure the ultimate in product safety.
Leistungsmerkmale und Vorteile
• PTFE faced (14B) styrene-butadiene rubber liner provides a totally inert inner seal and surface facing the sample or product•
Rechtliche Hinweise
WHEATON is a registered trademark of Wheaton Industries, Inc.