Z197130
3M™ disposable respirator
organic vapor/acid gas, size L
Alle Fotos(2)
About This Item
UNSPSC-Code:
46182002
NACRES:
NB.35
Empfohlene Produkte
Größe
L
Verpackung
pkg of 1 ea
Hersteller/Markenname
3M 5000
Allgemeine Beschreibung
Pre-assembled disposable respirators. Low-profile design provides improved comfort and less vision interference. Approved for: Organic vapor/acid gas and dusts/mists having a TWA not less than 0.05 mg/m3 or 2 mppcf.
NIOSH approved.
Protection factor: Up to 10 × PEL.
NIOSH approved.
Protection factor: Up to 10 × PEL.
Rechtliche Hinweise
3M is a trademark of 3M Company
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?