Merck
WHA420400
Whatman® plastic filter holders
Swin-Lok Holder, diam. 47 mm
Synonym(e):
Whatman filter, Whatman filters, Z742694
About This Item
UNSPSC-Code:
41104922
NACRES:
NB.24
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
polycarbonate housing
Sterilität
non-sterile
Leistungsmerkmale
Swin-Lok Holder
Verpackung
pkg of 8 ea
Hersteller/Markenname
Whatman 420400
Whatman Article No. 28419556 (US reference)
Durchmesser
47 mm
Allgemeine Beschreibung
Filter holders designed for microfiltration and ultra cleaning of small volumes of liquids using positive pressure.
Leistungsmerkmale und Vorteile
  • Designed for microfiltration and ultra cleaning of small volumes of liquids using positive pressure.
  • All three holders will accommodate Nucleopore track-etched and cast membranes.
  • Syringe compatible.
Sonstige Hinweise
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)