Merck
WHA10370392
Whatman® glass microfiber filters with organic binder, Grade GF 10
roll, W × L 35 mm × 30 m, pack of 1
Synonym(e):
fiber filters, glass filter, whatman filter
About This Item
UNSPSC-Code:
40161507
NACRES:
NB.24
Materialien
glass membrane (with organic binder)
roll
Sterilität
non-sterile
Verpackung
pack of 1
Hersteller/Markenname
Whatman 10370392
Whatman Article No. 28414800 (US reference)
B × L
35 mm × 30 m
Allgemeine Beschreibung
This filter with extreme mechanical stability and temperature resistant up to 180°C is used as a weighing aid for infrared weighing and as a roll filter in automatic air filtration units.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)