Merck
TEM-7596407
Sigma-Aldrich
OMNI Probe Copper 3-Post Lift-Out Grid
pack of 100 (per vial)
Synonym(e):
Omniprobe Lift-Out Grid, 3-Post, Side Access, CU
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Verpackung
pack of 100 (per vial)
Allgemeine Beschreibung
3 post copper lift-out grids, 35 micron thick with 1 edge lower for easy access.
Verpackung
Packaged in glass vials.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)