Merck
SYNM290004
Synthware all-glass vacuum/inert gas manifold with solid high vacuum stopcocks
number of ports, 4, Hose Connections: Front-left-right, Rear-right
Synonym(e):
Glass vacuum manifold, High vacuum manifold, Inert gas system, Synthware M290004, vacuum manifold, vacuum unit
Größe auswählen
1 EA
€ 750,00
€ 750,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 750,00
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
€ 750,00
Check Cart for Availability
Leistungsmerkmale
number of ports 4
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware M290004
Allgemeine Beschreibung
With 4 mm double oblique, high vacuum solid glass stopcocks precision ground with corresponding numbers on barrel and plug. Solid glass stopcocks have constant pressure retention devices. Beams are 22 mm O.D. with front-left-right and rear-right 10 mm hose connections. Distance between stopcocks is75 mm center to center.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)