Merck
SYNM150004
Synthware manifold with high vacuum stopcocks and #15 o-ring joints
Ports: 4
Synonym(e):
Synthware M150004, vacuum manifold, vacuum unit
Größe auswählen
1 EA
€ 843,00
€ 843,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 843,00
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.77
€ 843,00
Check Cart for Availability
Hersteller/Markenname
M150004
Allgemeine Beschreibung
Synthware glass manifold featuring 4 mm double hollow high vacuum stopcocks, this four-port manifold is fitted with two 14/20 and two 24/40 inner joints, with a 100 mm center-to-center distance between the joints. The #15 O-ring joint stopper (SYNS480015) and the #15 O-ring joint to serrated hose adapter must be ordered separately.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)