Merck
SYNF794000
Synthware modified evaporation flask
capacity 1000 mL, neck joint: ST/NS 24/40
Größe auswählen
1 EA
€ 111,00
€ 111,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 111,00
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
€ 111,00
Check Cart for Availability
Materialien
glass flask
round bottom flask
Verbindung
joint: ST/NS neck24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F794000
Kapazität
1000 mL
Allgemeine Beschreibung
Low profile design sits upright on bench and also provides for easy product access with a spatula. The unique flask shape is excellent for the controlled evaporation of foaming solvents such as toluene or xylene on a rotary evaporator
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)