Merck
SYNF464500
Synthware storage flask with standard taper side arm
capacity 500 mL, joint: ST/NS 24/40, valve size 0-4 mm
Synonym(e):
Synthware F464500, round flask with side arm
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
borosilicate glass flask
round bottom
single-neck
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F464500
Kapazität
500 mL
Ventilgröße
0-4 mm
Allgemeine Beschreibung
Supplied with high vacuum valve with PTFE O-ring at tip.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.