Merck
SYND441405
Synthware Dean Stark distillation receiver with vacuum jacketed uptake tube
5 mL, joint: ST/NS 14/20
Synonym(e):
Synthware D441405, Dean-Stark apparatus, Distilling receiver, Dean-Stark trap
Größe auswählen
1 EA
€ 209,00
€ 209,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 209,00
About This Item
UNSPSC-Code:
41104812
NACRES:
NB.51
€ 209,00
Check Cart for Availability
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware D441405
Kapazität
5 mL
Allgemeine Beschreibung
Similar to D451414 above but with the uptake tube vacuum jacketed to improve distillation performance. The jacket eliminates the possibility of condensation or recrystallization of distillate in the sidearm. With 2 mm PTFE stopcock.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)