Merck
SYNC368030
Synthware chromatography column with PTFE stopcock
I.D. 73.0 mm, L 305 mm
Synonym(e):
Synthware C368030, glass chromatograpy column
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Qualität
for chromatography
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware C368030
ID
73.0 mm
Länge
305 mm
Allgemeine Beschreibung
With reinforced beaded top. All Synthware columns are heavy wall designed to increase safety.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.