Merck
SYNA851922
Synthware U-shaped drying tube
joint: ST/NS 19/22
Synonym(e):
Synthware A851922, Drying tube adapter
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Materialien
colorless tube
glass tube
Verbindung
joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A851922
Allgemeine Beschreibung
With joint. Cotton is usually used to contain drying agent.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.