Merck
SYNA521922
Synthware inlet or thermometer adapter with compression cap
joint: ST/NS 19/22, O.D. 5.0-7.5 mm
Synonym(e):
Synthware A521922, thermometer inlet adapter
Größe auswählen
1 EA
€ 28,30
€ 28,30
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 28,30
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
€ 28,30
Check Cart for Availability
Verbindung
joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A521922
AD
5.0-7.5 mm
Allgemeine Beschreibung
With a compression cap and Viton® O-ring to form a vacuum tight seal. To use it as a syringe port, simply replace the O-ring with PTFE -lined septum. Cap accommodates 5.0 - 7.5mm O.D. thermometer. Supplied with one Nylon cap and one Viton® O-ring.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)