Merck
SYNA342440
Synthware twin hose connecting adapter
joint: ST/NS 24/40
Synonym(e):
Synthware A342440, Adapter, twin connecting hose
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A342440
Allgemeine Beschreibung
Adapter has two hose connections 180° from one another to permit a flow of inert gas over the apparatus.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.