Merck
MT30014098
Mettler Toledo flow-cell for InLab® trace conductivity probe
Größe auswählen
1 EA
€ 1.390,00
€ 1.390,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 1.390,00
About This Item
UNSPSC-Code:
41115600
NACRES:
NB.79
€ 1.390,00
Check Cart for Availability
Hersteller/Markenname
Mettler-Toledo 30014098
Allgemeine Beschreibung
Using the Flow Cell the InLab® Trace the highest repeatability and reproducibility ever can be achieved.
The Flow Cell InLab® Trace was built at an angle to prevent air bubbles being trapped. Your workflow is optimized to the max. Enjoy the peace of mind.
Rechtliche Hinweise
InLab is a registered trademark of Mettler-Toledo, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)