Merck
CLS5720100
Pyrex® volumetric sugar analysis flask
capacity 100 mL
Synonym(e):
flasks glass, volumetric flasks, volumetric glass, Corning® 5720
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Hersteller/Markenname
Corning 5720-100
tol. measuring range
0.08 mL
Kapazität
100 mL
Allgemeine Beschreibung
With a short neck and a slightly flared top. For the preparation of sugar solutions for polarization tests. Conforms to ASTM E-694.Reference: Sugar Division of the National Bureau of Standards.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)