Merck
CAD4021
Standard point hypodermic needles, 304 stainless steel
gauge 25, L 1 in., nickel plated hub
Größe auswählen
12 EA
€ 473,00
€ 473,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
12 EA
€ 473,00
About This Item
UNSPSC-Code:
42142523
NACRES:
NB.03
€ 473,00
Check Cart for Availability
Materialien
304 stainless steel needle
nickel plated hub
Verpackung
pack of 12 ea
Hersteller/Markenname
Cadence Science 4021
Länge
1 in.
Manometer
25
Verwandte Kategorien
Allgemeine Beschreibung
Hypodermic needles are precision made and designed to ensure strict adherence with U.S. Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannula are fabricated of type 304 stainless steel tubing which must pass rigorous quality control prior to acceptance and use.
Standard nickle plated Luer hubs comply with Luer taper specifications ISO 594. This assures an interchangeable fit mating with Luer slips and Luer locks. Contoured hub cavity facilitates cleaning.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)