Merck
BR81919
BRAND® burette straight stopcock
for burette capacity 50 mL, glass key
Synonym(e):
Accessories, Burette stopcock, Burettes, Stopcocks
About This Item
UNSPSC-Code:
41121808
NACRES:
NB.43
Materialien
amber
glass key
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 81919
Allgemeine Beschreibung
BRAND burettes are always calibrated to deliver (TD, Ex).
Waiting times:
BLAUBRAND® - waiting time 30 sec. SILBERBRAND - no waiting time
A waiting time is usually not necessary if a titration is performed (see DIN EN ISO 385).
All BLAUBRAND® burettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
Rechtliche Hinweise
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)