Merck
BR781530
Brand®1.2 ML Tubes
individual caps, PE
Synonym(e):
Accessories, Caps, Sample Storage, Tube racks, Tubes
Größe auswählen
960 EA
€ 70,20
€ 70,20
Check Cart for Availability
Größe auswählen
Ansicht ändern
960 EA
€ 70,20
About This Item
UNSPSC-Code:
41122804
NACRES:
NB.22
€ 70,20
Check Cart for Availability
Materialien
PE
Sterilität
non-sterile
Leistungsmerkmale
individual caps
Verpackung
pack of 960 pieces (bulk packed)
Hersteller/Markenname
BRAND 781530
Kapazität
1.2 mL
Allgemeine Beschreibung
PE. Caps are free from RNase, DNase and DNA and not autoclavable. Ideal for biological tests, such as PCR, cell uptake studies, RIA and EIA. Designed for storing and freezing down to -80 °C, and for transporting reagents and samples. Compatible with standard microplates and suitable for working with multichannel pipettes. (Size of tubes: outer Ø 8.8 mm, height 45 mm.)BIO-CERT® CERTIFIED QUALITY: Free of human DNA, RNase, DNase and Pyrogens
Rechtliche Hinweise
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)