Merck
BR458732
BRAND® filter crucible
borosilicate glass, capacity 50 mL, 2 D 2
Synonym(e):
Filter, Filter crucible
About This Item
UNSPSC-Code:
41122200
NACRES:
NB.43
Materialien
borosilicate glass
Hersteller/Markenname
BRAND 458732
Kapazität
50 mL
Porengröße
40-100 μm
Allgemeine Beschreibung
The BRAND® Filter Crucible made from Boro 3.3 glass is an essential tool for efficient filtration and separation processes in laboratory applications.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG