Merck
BR41648
BRAND® graduated cylinder low form embossed scale
volume 250 mL, polypropylene
Synonym(e):
Graduated Cylinders, Plastics
Größe auswählen
5 EA
€ 33,50
€ 33,50
Check Cart for Availability
Größe auswählen
Ansicht ändern
5 EA
€ 33,50
About This Item
UNSPSC-Code:
41121805
NACRES:
NB.43
€ 33,50
Check Cart for Availability
Materialien
polypropylene
Hersteller/Markenname
BRAND 41648
Volumen
250 mL
Allgemeine Beschreibung
The BRAND® Graduated Cylinder, Low Form with Embossed Scale, is designed for precise liquid measurement in laboratory settings. Made from high-clarity polypropylene (PP), this cylinder is calibrated to contain (TC, In), ensuring accurate volume readings. Its low form design enhances stability and ease of use, while the embossed scale provides clear visibility for quick reference. This graduated cylinder can withstand exposure to temperatures up to 80 °C without exceeding permanent error limits, making it suitable for a variety of applications. Reliable and durable, this graduated cylinder is an essential tool for any laboratory.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)