Merck
BAF371090033
Spinbar® Magnetrührstäbchen
octagonal blue PTFE-coated, L × diam. 3 in. (76.2 mm) × 0.5 in. (12.7 mm)
Synonym(e):
Spinbar® magnetic stir bar
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.51
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
octagonal blue PTFE-coated
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art F37109-0033
L × Durchm.
3 in. (76.2 mm) × 0.5 in. (12.7 mm)
Allgemeine Beschreibung
Mit PTFE-Überzug. Mit starken Permanentmagneten aus Alnico V. Achteckiger Querschnitt, mit Drehring. Ovale Rührfische sind besonders für Gefäße mit rundem Boden geeignet.
  • Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
  • Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
  • Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
  • Autoclave safe.
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape. Choose from red, yellow or blue.
Leistungsmerkmale und Vorteile
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE coating – Superior Chemical Compatibility
  • Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
  • Pivot ring aids in reducing friction and chattering
  • Autoclavable
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Spinbar is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)