Merck
383287
Sigma-Aldrich
Zirkoniumsilicat
−325 mesh
Synonym(e):
Zirconium(IV)-siliciumoxid
About This Item
Lineare Formel:
ZrSiO4
CAS-Nummer:
Molekulargewicht:
183.31
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12352103
PubChem Substanz-ID:
NACRES:
NA.23
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Form
powder
Eignung der Reaktion
core: zirconium
Partikelgröße
−325 mesh
SMILES String
[Zr+4].[O-][Si]([O-])([O-])[O-]
InChI
1S/O4Si.Zr/c1-5(2,3)4;/q-4;+4
InChIKey
GFQYVLUOOAAOGM-UHFFFAOYSA-N
Leistungsmerkmale und Vorteile
Often cited as a potential alternative gate dielectric to silica.
Lagerklassenschlüssel
11 - Combustible Solids
WGK
WGK 3
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Persönliche Schutzausrüstung
dust mask type N95 (US), Eyeshields, Gloves
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
N Fathabadi et al.
Radiation protection dosimetry, 145(4), 400-404 (2010-12-15)
Zircon contains small amounts of uranium, thorium and radium in its crystalline structure. The ceramic industry is one of the major consumers of zirconium compounds that are used as an ingredient at ∼10-20 % by weight in glaze. In this
E K H Salje et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 23(10), 105402-105402 (2011-02-23)
Iron is a ubiquitous impurity in metamict (radiation-damaged and partially amorphized) materials such as titanite (CaSiTiO(5)). Using (57)Fe Mössbauer spectroscopy we find that iron in metamict titanite is partitioned between amorphous and crystalline regions based on valence. Trivalent iron exists
Lubica Hallmann et al.
Journal of dentistry, 40(9), 723-735 (2012-05-23)
The aim of this study was to characterize the surface of Y-TZP after abrasion with various airborne particles. The Y-TZP blanks were cut into 44 discs and sintered according to the manufacturer's instructions. The specimens were treated as follows: (a)
Terrence J Blackburn et al.
Science (New York, N.Y.), 340(6135), 941-945 (2013-03-23)
The end-Triassic extinction is characterized by major losses in both terrestrial and marine diversity, setting the stage for dinosaurs to dominate Earth for the next 136 million years. Despite the approximate coincidence between this extinction and flood basalt volcanism, existing
Selective accumulation of zircons and ilmenites in diverticula of the sea urchin Scaphechinus mirabilis (Agazzis, 1863).
Yu N Elkin et al.
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections, 446, 297-299 (2012-11-07)