Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G7757

Sigma-Aldrich

Glycerol

ReagentPlus®, ≥99.0% (GC)

Synonym(s):

1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21
technique(s):
electrophoresis: suitable
bp:
182 °C/20 mmHg (lit.)
vapor pressure:
<1 mmHg ( 20 °C)

vapor density

3.1 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99.0% (GC)

form

viscous liquid

autoignition temp.

698 °F

expl. lim.

2.6-11.3 %

technique(s)

electrophoresis: suitable

impurities

≤0.1% sulfated ash
≤1.0% water (Karl Fischer)

refractive index

n20/D 1.474 (lit.)

pH

5.5-8

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

density

1.25 g/mL (lit.)

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hovsep Mahdessian et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(24), 8913-8918 (2014-06-14)
Genome-wide association studies have identified a locus on chromosome 19 associated with plasma triglyceride (TG) concentration and nonalcoholic fatty liver disease. However, the identity and functional role of the gene(s) responsible for these associations remain unknown. Of 19 expressed genes
Shelley L Forrest et al.
The Journal of comparative neurology, 522(17), 3900-3927 (2014-07-22)
Artemin is a member of the glial cell line-derived neurotrophic factor (GDNF) family that has been strongly implicated in development and regeneration of autonomic nerves and modulation of nociception. Whereas other members of this family (GDNF and neurturin) primarily target
Marc Simard et al.
The Journal of clinical endocrinology and metabolism, 100(1), E129-E139 (2014-10-17)
Corticosteroid-binding globulin (CBG) is encoded by SERPINA6. It is the major plasma binding protein of glucocorticoids and regulates plasma cortisol levels and bioavailability in humans. Several proteases target CBG and disrupt its steroid-binding properties. To date, most genetic deficiencies that
Nazish Ahmed et al.
Tissue engineering. Part A, 20(15-16), 2224-2233 (2014-03-13)
One of the factors preventing clinical application of regenerative medicine to degenerative cartilage diseases is a suitable source of cells. Chondrocytes, the only cell type of cartilage, grown in vitro under culture conditions to expand cell numbers lose their phenotype
T W Chew et al.
Oncogene, 33(21), 2717-2727 (2013-07-03)
RAS and Rho small GTPases are key molecular switches that control cell dynamics, cell growth and tissue development through their distinct signaling pathways. Although much has been learnt about their individual functions in both cell and animal models, the physiological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service